Skip to Content
Merck
4-Ethoxy-3-nitrobenzaldehyde 97%

4-Ethoxy-3-nitrobenzaldehyde

4-Ethoxy-3-nitrobenzaldehyde 97%
Empirical Formula (Hill Notation):
C9H9NO4
CAS Number:
Molecular Weight:
195.17

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing