Skip to Content
Merck
4-Ethoxy-3-formylphenylboronic acid ≥95%

4-Ethoxy-3-formylphenylboronic acid

4-Ethoxy-3-formylphenylboronic acid ≥95%
Linear Formula:
C6H3OCH2CH3CHOB(OH)2
CAS Number:
Molecular Weight:
193.99

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing