Skip to Content
Merck
4-Chloro-3-iodo-pyridine 97%

4-Chloro-3-iodo-pyridine

4-Chloro-3-iodo-pyridine 97%
Empirical Formula (Hill Notation):
C5H3ClIN
CAS Number:
Molecular Weight:
239.44

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing