Skip to Content
Merck
4-(Bromomethyl)phenylacetic acid 95%

4-(Bromomethyl)phenylacetic acid

4-(Bromomethyl)phenylacetic acid 95%
Linear Formula:
BrCH2C6H4CH2CO2H
CAS Number:
Molecular Weight:
229.07

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing