Skip to Content
Merck
4-Bromoisoindoline hydrochloride 95%

4-Bromoisoindoline hydrochloride

4-Bromoisoindoline hydrochloride 95%
Synonym(s):
4-Bromo-1H-isoindoline hydrochloride
Empirical Formula (Hill Notation):
C8H8BrN · HCl
CAS Number:
Molecular Weight:
234.52

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing