Skip to Content
Merck
4-Bromobenzyl methyl ether 97%

4-Bromobenzyl methyl ether

4-Bromobenzyl methyl ether 97%
Linear Formula:
BrC6H4CH2OCH3
CAS Number:
Molecular Weight:
201.06

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing