Skip to Content
Merck
4-Bromo-3-(trifluoromethyl)pyridine hydrobromide 97%

4-Bromo-3-(trifluoromethyl)pyridine hydrobromide

4-Bromo-3-(trifluoromethyl)pyridine hydrobromide 97%
Empirical Formula (Hill Notation):
C6H4Br2F3N
CAS Number:
Molecular Weight:
306.91

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing