Skip to Content
Merck
4-Bromo-2-methylphenol 97%

4-Bromo-2-methylphenol

4-Bromo-2-methylphenol 97%
Synonym(s):
4-Bromo-o-cresol
Empirical Formula (Hill Notation):
C7H7BrO
CAS Number:
Molecular Weight:
187.03

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing