Skip to Content
Merck
4-Bromo-1,3,5-trimethylpyrazole 98%

4-Bromo-1,3,5-trimethylpyrazole

4-Bromo-1,3,5-trimethylpyrazole 98%
Empirical Formula (Hill Notation):
C6H9BrN2
CAS Number:
Molecular Weight:
189.05

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing