
4arm-PEG10K-Succinimidyl Succinate

Synonym(s):
4arm-PEG-Succinimidyl Succinate
Linear Formula:
C(CH2O(CH2CH2O)nCOCH2CH2COOC4H4O2N)4
Products
Brand
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing