Skip to Content
Merck
4-Amino-3-nitrobenzonitrile 98%

4-Amino-3-nitrobenzonitrile

4-Amino-3-nitrobenzonitrile 98%
Linear Formula:
H2NC6H3(NO2)CN
CAS Number:
Molecular Weight:
163.13

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing