Skip to Content
Merck
4-Amino-3-iodopyridine 96%

4-Amino-3-iodopyridine

4-Amino-3-iodopyridine 96%
Synonym(s):
3-Iodo-4-aminopyridine
Empirical Formula (Hill Notation):
C5H5IN2
CAS Number:
Molecular Weight:
220.01

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing