4-Amidinophenylmethanesulfonyl fluoride hydrochloride
Synonym(s):
4-Amidinobenzylsulfonyl fluoride hydrochloride, p-APMSF
Empirical Formula (Hill Notation):
C8H9FN2O2S · HCl
CAS Number:
Molecular Weight:
252.69
Products
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing