Skip to Content
Merck
4-Acetylbiphenyl 98%

4-Acetylbiphenyl

4-Acetylbiphenyl 98%
Synonym(s):
4-Biphenyl methyl ketone, 4-Phenylacetophenone
Linear Formula:
C6H5C6H4COCH3
CAS Number:
Molecular Weight:
196.24
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing