Skip to Content
Merck
4,6-Dichloro-5-methylpyrimidine 97%

4,6-Dichloro-5-methylpyrimidine

4,6-Dichloro-5-methylpyrimidine 97%
Empirical Formula (Hill Notation):
C5H4Cl2N2
CAS Number:
Molecular Weight:
163.00

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing