Skip to Content
Merck
4,5-Difluoro-2-nitroaniline 98%

4,5-Difluoro-2-nitroaniline

4,5-Difluoro-2-nitroaniline 98%
Linear Formula:
F2C6H2(NO2)NH2
CAS Number:
Molecular Weight:
174.10

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing