Skip to Content
Merck
4,4-Dimethyl-2-phenyl-2-oxazoline 96%

4,4-Dimethyl-2-phenyl-2-oxazoline

4,4-Dimethyl-2-phenyl-2-oxazoline 96%
Empirical Formula (Hill Notation):
C11H13NO
CAS Number:
Molecular Weight:
175.23

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing