Skip to Content
Merck
4,4′-Dimethoxybenzoin 95%

4,4′-Dimethoxybenzoin

4,4′-Dimethoxybenzoin 95%
Synonym(s):
2-Hydroxy-4′-methoxy-2-(4-methoxyphenyl)acetophenone, p-Anisoin
Linear Formula:
4-(CH3O)C6H4CH(OH)COC6H4-4-(OCH3)
CAS Number:
Molecular Weight:
272.30
EC Number:

Products

Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing