
4,4′-Diisothiocyanatostilbene-2,2′-disulfonic acid disodium salt hydrate

Synonym(s):
DIDS, Disodium 4,4′-diisothiocyanatostilbene-2,2′-disulfonate
Empirical Formula (Hill Notation):
C16H8N2Na2O6S4 · xH2O
CAS Number:
Molecular Weight:
498.48 (anhydrous basis)
Products
Brand
Color
Formula Weight
Physical Form
Purity
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing