![4,4′,4′′-Tris[2-naphthyl(phenyl)amino]triphenylamine 97%](/deepweb/assets/sigmaaldrich/product/structures/699/220/b795fd33-c46d-43c7-843c-26704abdec45/640/b795fd33-c46d-43c7-843c-26704abdec45.png)
4,4′,4′′-Tris[2-naphthyl(phenyl)amino]triphenylamine
![4,4′,4′′-Tris[2-naphthyl(phenyl)amino]triphenylamine 97%](/deepweb/assets/sigmaaldrich/product/structures/699/220/b795fd33-c46d-43c7-843c-26704abdec45/640/b795fd33-c46d-43c7-843c-26704abdec45.png)
Synonym(s):
2-TNATA, 2TNATA
Empirical Formula (Hill Notation):
C66H48N4
CAS Number:
Molecular Weight:
897.11
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing