Skip to Content
Merck
3-Methoxy-4-methylaniline 98%

3-Methoxy-4-methylaniline

3-Methoxy-4-methylaniline 98%
Synonym(s):
4-Methyl-m-anisidine, o-Cresidine
Empirical Formula (Hill Notation):
C8H11NO
CAS Number:
Molecular Weight:
137.18
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing