Skip to Content
Merck
3-Iodo-4-methylaniline 97%

3-Iodo-4-methylaniline

3-Iodo-4-methylaniline 97%
Linear Formula:
IC6H3(CH3)NH2
CAS Number:
Molecular Weight:
233.05
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing