Skip to Content
Merck
3-Ethyl-2-thioxo-4-oxazolidinone 97%

3-Ethyl-2-thioxo-4-oxazolidinone

3-Ethyl-2-thioxo-4-oxazolidinone 97%
Empirical Formula (Hill Notation):
C5H7NO2S
CAS Number:
Molecular Weight:
145.18
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing