
3-Cyano-7-ethoxycoumarin

Empirical Formula (Hill Notation):
C12H9NO3
CAS Number:
Molecular Weight:
215.20
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing