Skip to Content
Merck
3-Chloro-2,4,5,6-tetrafluoropyridine 98%

3-Chloro-2,4,5,6-tetrafluoropyridine

3-Chloro-2,4,5,6-tetrafluoropyridine 98%
Empirical Formula (Hill Notation):
C5ClF4N
CAS Number:
Molecular Weight:
185.51

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing