Skip to Content
Merck
3-Aminobenzamidine dihydrochloride 96%

3-Aminobenzamidine dihydrochloride

3-Aminobenzamidine dihydrochloride 96%
Linear Formula:
H2NC6H4C(=NH)NH2·2HCl
CAS Number:
Molecular Weight:
208.09
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing