Skip to Content
Merck
3-Amino-5-methylpyrazole 97%

3-Amino-5-methylpyrazole

3-Amino-5-methylpyrazole 97%
Synonym(s):
5-Methyl-3-pyrazolamine
Empirical Formula (Hill Notation):
C4H7N3
CAS Number:
Molecular Weight:
97.12

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing