Skip to Content
Merck
3,7-Dimethyluric acid ≥95.0% (HPLC)

3,7-Dimethyluric acid

3,7-Dimethyluric acid ≥95.0% (HPLC)
Synonym(s):
3,7-Dimethyl-2,6,8-trihydroxypurine
Empirical Formula (Hill Notation):
C7H8N4O3
CAS Number:
Molecular Weight:
196.16

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing