
3,5-Dimethyl-4-methoxybenzoic acid

Synonym(s):
3,5-Dimethyl-p-anisic acid
Empirical Formula (Hill Notation):
C10H12O3
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing