Skip to Content
Merck
3,5-Dibenzyloxyacetophenone 98%

3,5-Dibenzyloxyacetophenone

3,5-Dibenzyloxyacetophenone 98%
Linear Formula:
(C6H5CH2O)2C6H3COCH3
CAS Number:
Molecular Weight:
332.39
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing