Skip to Content
Merck
3,4-Dimethoxythiophenol 97%

3,4-Dimethoxythiophenol

3,4-Dimethoxythiophenol 97%
Synonym(s):
3,4-Dimethoxybenzenethiol
Empirical Formula (Hill Notation):
C8H10O2S
CAS Number:
Molecular Weight:
170.23

Products

Product Category
Brand
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing