Skip to Content
Merck
3,4-Dimethoxy-N-methylphenethylamine hydrochloride 98%

3,4-Dimethoxy-N-methylphenethylamine hydrochloride

3,4-Dimethoxy-N-methylphenethylamine hydrochloride 98%
Synonym(s):
N-Methylhomoveratrylamine hydrochloride
Linear Formula:
(CH3O)2C6H3CH2CH2NHCH3·HCl
CAS Number:
Molecular Weight:
231.72
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing