Skip to Content
Merck
3,4,5-Trifluorophenol ≥98.0%

3,4,5-Trifluorophenol

3,4,5-Trifluorophenol ≥98.0%
Empirical Formula (Hill Notation):
C6H3F3O
CAS Number:
Molecular Weight:
148.08
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing