Skip to Content
Merck
2-Methoxyethyl acetoacetate Arxada quality, ≥97.5% (GC)

2-Methoxyethyl acetoacetate

2-Methoxyethyl acetoacetate Arxada quality, ≥97.5% (GC)
Linear Formula:
CH3COCH2CO2CH2CH2OCH3
CAS Number:
Molecular Weight:
160.17

Products

Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Manufacturer
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing