Skip to Content
Merck
2-methoxy-6-(tributylstannyl)pyridine 96%

2-methoxy-6-(tributylstannyl)pyridine

2-methoxy-6-(tributylstannyl)pyridine 96%
Empirical Formula (Hill Notation):
C18H33NOSn
Molecular Weight:
398.17

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing