Skip to Content
Merck
2-Isobutyl-4-methyl-1,3-dioxolane mixture of isomers, ≥97%

2-Isobutyl-4-methyl-1,3-dioxolane

2-Isobutyl-4-methyl-1,3-dioxolane mixture of isomers, ≥97%
Empirical Formula (Hill Notation):
C8H16O2
CAS Number:
Molecular Weight:
144.21
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Showing 1-1 of 1
Compare
Product Number
Description
Pricing