Skip to Content
Merck
2-Iodofluorene 98%

2-Iodofluorene

2-Iodofluorene 98%
Synonym(s):
2-Iodo-9H-fluorene
Empirical Formula (Hill Notation):
C13H9I
CAS Number:
Molecular Weight:
292.12

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing