
2-Hydroxy-propoxycarbazone

Synonym(s):
2-{{{[4,5-Dihydro-3-(2-hydroxypropoxy)-4-methyl-5-oxo-1H-1,2,4-triazol-1-yl]carbonyl}amino}sulfonyl}benzoic acid methyl ester
Empirical Formula (Hill Notation):
C15H18N4O8S
CAS Number:
Molecular Weight:
414.39
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing