Skip to Content
Merck
2-Hydroxy-4-methyl-3-nitropyridine 98%

2-Hydroxy-4-methyl-3-nitropyridine

2-Hydroxy-4-methyl-3-nitropyridine 98%
Synonym(s):
4-Methyl-3-nitro-2-pyridone
Empirical Formula (Hill Notation):
C6H6N2O3
CAS Number:
Molecular Weight:
154.12

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing