Skip to Content
Merck
2-Fluoro-5-nitrobenzaldehyde 97%

2-Fluoro-5-nitrobenzaldehyde

2-Fluoro-5-nitrobenzaldehyde 97%
Linear Formula:
FC6H3(NO2)CHO
CAS Number:
Molecular Weight:
169.11

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing