
2-(Diisopropylphosphino)ethylamine solution

Synonym(s):
2-(Diisopropylphosphino)ethylamine, 2-[Bis(1-methylethyl)phosphino]ethanamine
Empirical Formula (Hill Notation):
C8H20NP
CAS Number:
Molecular Weight:
161.22
Products
Brand
Formula Weight
Physical Form
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing