Skip to Content
Merck
2-Chlorobenzyl alcohol 99%

2-Chlorobenzyl alcohol

2-Chlorobenzyl alcohol 99%
Linear Formula:
ClC6H4CH2OH
CAS Number:
Molecular Weight:
142.58
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing