Skip to Content
Merck
2-Chloro-6-methylpyridine 99%

2-Chloro-6-methylpyridine

2-Chloro-6-methylpyridine 99%
Synonym(s):
6-Chloro-2-picoline
Empirical Formula (Hill Notation):
C6H6ClN
CAS Number:
Molecular Weight:
127.57
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing