
2-Bromophenylacetonitrile

Synonym(s):
2-Bromobenzyl cyanide
Linear Formula:
BrC6H4CH2CN
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing