Skip to Content
Merck
2-Bromo-6-nitrophenol 97%

2-Bromo-6-nitrophenol

2-Bromo-6-nitrophenol 97%
Empirical Formula (Hill Notation):
C6H4BrNO3
CAS Number:
Molecular Weight:
218.00

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing