
2-Acetamido-2-deoxy-D-glucono-1,5-lactone

Synonym(s):
2-(Acetylamino)-2-deoxy-D-gluconic acid δ-lactone, 2-Acetamido-2-deoxy-D-glucono-δ-lactone, N-Acetylglucosamino-1,5-lactone
Empirical Formula (Hill Notation):
C8H13NO6
CAS Number:
Molecular Weight:
219.19
Products
Product Category
Brand
Formula Weight
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing