Skip to Content
Merck
2-Acetamido-1-N-(β-L-aspartyl)-2-deoxy-β-D-glucopyranosylamine ≥98%

2-Acetamido-1-N-(β-L-aspartyl)-2-deoxy-β-D-glucopyranosylamine

2-Acetamido-1-N-(β-L-aspartyl)-2-deoxy-β-D-glucopyranosylamine ≥98%
Synonym(s):
β-D-GlcNAc-(1→N)-Asn, 2-Acetamido-1-β-(L-aspartamido)-1,2-dideoxy-D-glucose, L-Asparagine, N-[2-(acetylamino)-2-deoxy-beta-D-glucopyranosyl]
Empirical Formula (Hill Notation):
C12H21N3O8
CAS Number:
Molecular Weight:
335.31

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing