Skip to Content
Merck
2,5-Dimethoxyphenethylamine 97%

2,5-Dimethoxyphenethylamine

2,5-Dimethoxyphenethylamine 97%
Linear Formula:
(CH3O)2C6H3CH2CH2NH2
CAS Number:
Molecular Weight:
181.23

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing