2,5-Dibromo-3-methylpyridine
Empirical Formula (Hill Notation):
C6H5Br2N
CAS Number:
Molecular Weight:
250.92
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing