![2-[4-(Dimethylamino)phenyl]ethanol ≥99%](/deepweb/assets/sigmaaldrich/product/structures/741/460/ce355b70-7460-4ef9-920b-19b737bae725/640/ce355b70-7460-4ef9-920b-19b737bae725.png)
2-[4-(Dimethylamino)phenyl]ethanol
![2-[4-(Dimethylamino)phenyl]ethanol ≥99%](/deepweb/assets/sigmaaldrich/product/structures/741/460/ce355b70-7460-4ef9-920b-19b737bae725/640/ce355b70-7460-4ef9-920b-19b737bae725.png)
Synonym(s):
4-(Dimethylamino)phenethyl alcohol
Linear Formula:
(CH3)2NC6H4CH2CH2OH
CAS Number:
Molecular Weight:
165.23
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing